The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001285439.1 | NP_001272368.1 | 15 - 16 | 1334 | 81 |
NM_001285440.1 | NP_001272369.1 | 15 - 16 | 1332 | 81 | |
NM_152756.4 | NP_689969.2 | 15 - 16 | 1334 | 81 | |
XM_006714463.3 | XP_006714526.1 | 15 - 16 | 1347 | 81 | |
XM_011514005.2 | XP_011512307.1 | 15 - 16 | 1347 | 81 | |
XM_011514006.2 | XP_011512308.1 | 14 - 15 | 1151 | 81 | |
XM_017009311.1 | XP_016864800.1 | 14 - 15 | 1267 | 81 | |
XM_017009312.1 | XP_016864801.1 | 14 - 15 | 1267 | 81 | |
XM_017009313.1 | XP_016864802.1 | 14 - 15 | 1288 | 81 | |
XM_017009314.1 | XP_016864803.1 | 14 - 15 | 1257 | 81 | |
XM_017009315.1 | XP_016864804.1 | 14 - 15 | 1257 | 81 | |
GenBank mRNA | AY515854.1 | - | 15 - 16 | 1330 | 81 |
BC137163.1 | - | 15 - 16 | 1327 | 81 | |
BC137164.1 | - | 15 - 16 | 1327 | 81 | |
BC144509.1 | - | 15 - 16 | 1327 | 81 | |
CR749280.1 | - | 15 - 16 | 1313 | 81 |