The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003061.2 | NP_003052.2 | 8 - 9 | 1047 | 108 |
GenBank mRNA | AB011537.3 | - | 8 - 9 | 1047 | 108 |
AB017167.1 | - | 8 - 9 | 1027 | 108 | |
AY029183.1 | - | 8 - 9 | 744 | 108 | |
BC146761.1 | - | 8 - 9 | 1047 | 108 | |
BC146851.1 | - | 8 - 9 | 960 | 108 |