Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GNB3
Entrez Gene ID
2784
Gene Name
G protein subunit beta 3
Gene Aliases
CSNB1H
Chromosome Location
Chr.12: 6840854 - 6847400 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001297571.1 NP_001284500.1 6 - 7 635 83
NM_002075.3 NP_002066.1 6 - 7 635 83
XM_011520953.2 XP_011519255.1 6 - 7 567 83
GenBank mRNA AB209283.1 - 5 - 6 1776 83
AB593098.1 - 6 - 7 561 83
AB593099.1 - 6 - 7 560 83
AF501884.1 - 5 - 6 425 83
AK313091.1 - 6 - 7 515 83
BC000115.1 - 6 - 7 548 83
BC002454.2 - 6 - 7 555 83
BC015920.1 - 6 - 7 514 83
BT009800.1 - 5 - 6 425 83
M31328.1 - 5 - 6 431 83

View Full Product Details