The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040194.1 | NP_001035284.1 | 3 - 4 | 299 | 56 |
NM_001040195.1 | NP_001035285.1 | 2 - 3 | 264 | 56 | |
NM_001040196.1 | NP_001035286.1 | 4 - 5 | 396 | 56 | |
NM_001040197.1 | NP_001035287.1 | 4 - 5 | 396 | 56 | |
NM_020350.4 | NP_065083.3 | 3 - 4 | 299 | 56 | |
XM_011541799.2 | XP_011540101.2 | 4 - 5 | 313 | 56 | |
XM_011541800.2 | XP_011540102.1 | 4 - 5 | 313 | 56 | |
XM_011541802.1 | XP_011540104.1 | 5 - 6 | 410 | 56 | |
GenBank mRNA | AF312374.1 | - | 3 - 4 | 230 | 56 |
AK290042.1 | - | 3 - 4 | 242 | 56 | |
BC011946.1 | - | 3 - 4 | 205 | 56 | |
BC017328.2 | - | 3 - 4 | 237 | 56 | |
BE782705.1 | - | - | 349 | 56 | |
BI117521.1 | - | 3 - 4 | 271 | 56 | |
CA866314.1 | - | 2 - 3 | 270 | 56 | |
CD674964.1 | - | 4 - 5 | 276 | 56 |