The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040197.1 | NP_001035287.1 | 5 - 6 | 584 | 88 |
NM_020350.4 | NP_065083.3 | 4 - 5 | 487 | 88 | |
XM_011541800.2 | XP_011540102.1 | 5 - 6 | 501 | 88 | |
GenBank mRNA | AF165187.1 | - | 4 - 5 | 363 | 88 |
AF312374.1 | - | 4 - 5 | 418 | 88 | |
BI117521.1 | - | 4 - 5 | 459 | 88 | |
CD674964.1 | - | 5 - 6 | 464 | 88 |