The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000492.3 | NP_000483.3 | 14 - 15 | 2622 | 140 |
XM_011515751.2 | XP_011514053.1 | 15 - 16 | 2595 | 140 | |
XM_011515753.1 | XP_011514055.1 | 15 - 16 | 2712 | 140 | |
XM_011515754.2 | XP_011514056.1 | 17 - 18 | 3687 | 140 | |
XM_017011699.1 | XP_016867188.1 | 16 - 17 | 3433 | 140 |