Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- NAG20
- Entrez Gene ID
-
100129703
- Gene Name
- NAG20
- Gene Aliases
-
-
- Chromosome Location
-
-
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AF210649.1 |
- |
1 - 1 |
948 |
95 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
diacylglycerol kinase iota |
DGKI |
DGK-IOTA |
|
Sense |
Chr. - 137384283 - 137846864 on Build GRCh38 |
Downstream |
cAMP responsive element binding protein 3 like 2 |
CREB3L2 |
BBF2H7 |
206.2kb
|
Sense |
Chr. - 137874979 - 138002101 on Build GRCh38 |
View Full Product Details