The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033547.3 | NP_291025.3 | 17 - 18 | 2124 | 97 |
XM_011545352.2 | XP_011543654.1 | 18 - 19 | 2293 | 97 | |
XM_017018560.1 | XP_016874049.1 | 17 - 18 | 2157 | 97 | |
GenBank mRNA | AF172822.1 | - | 4 - 5 | 510 | 97 |
AK290866.1 | - | 17 - 18 | 2120 | 97 | |
AL834171.1 | - | 17 - 18 | 2067 | 97 | |
BC006369.1 | - | 5 - 6 | 493 | 97 | |
BC008013.1 | - | 13 - 14 | 1577 | 97 | |
BC009859.2 | - | 7 - 8 | 783 | 97 | |
BC015664.2 | - | 18 - 19 | 2434 | 97 |