Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GDAP1L1
Entrez Gene ID
78997
Gene Name
ganglioside induced differentiation associated protein 1 like 1
Gene Aliases
dJ881L22.1, dJ995J12.1.1
Chromosome Location
Chr.20: 44247099 - 44280917 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256737.1 NP_001243666.1 3 - 4 687 49
NM_001256738.1 NP_001243667.1 3 - 4 477 49
NM_024034.4 NP_076939.3 3 - 4 630 49
GenBank mRNA AF123764.1 - 3 - 4 529 49
AF123765.1 - 3 - 4 572 49
AF277566.1 - 3 - 4 585 49
AK056251.1 - 3 - 4 620 49
AK225166.1 - 3 - 4 620 49
AK299259.1 - 4 - 5 542 49
AK299700.1 - 3 - 4 636 49
BC000199.1 - 3 - 4 606 49
BC009014.1 - 3 - 4 569 49

View Full Product Details