The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007238.4 | NP_009169.3 | 1 - 2 | 229 | 96 |
NM_183397.2 | NP_899634.1 | 1 - 2 | 229 | 96 | |
GenBank mRNA | AF072864.1 | - | 1 - 2 | 151 | 96 |
AK023879.1 | - | 1 - 2 | 147 | 96 | |
AK297018.1 | - | 1 - 2 | 218 | 96 | |
AK301532.1 | - | 1 - 2 | 209 | 96 | |
AK316589.1 | - | 1 - 2 | 209 | 96 | |
AL136648.1 | - | 1 - 2 | 200 | 96 | |
BC001147.1 | - | 1 - 2 | 130 | 96 | |
BQ278083.1 | - | 1 - 2 | 197 | 96 | |
CR533471.1 | - | 1 - 2 | 106 | 96 | |
DB495125.1 | - | 1 - 2 | 229 | 96 |