Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- SDCBP2
- Entrez Gene ID
-
27111
- Gene Name
- syndecan binding protein 2
- Gene Aliases
-
SITAC, SITAC18, ST-2
- Chromosome Location
-
Chr.20: 1309909 - 1329235 on Build GRCh38
- Gene Symbol
- FKBP1A-SDCBP2
- Entrez Gene ID
-
100528031
- Gene Name
- FKBP1A-SDCBP2 readthrough (NMD candidate)
- Gene Aliases
-
-
- Chromosome Location
-
Chr.20: 1309909 - 1393172 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_037661.1 |
|
3 - 4 |
329 |
72 |
GenBank mRNA |
AK309026.1 |
- |
3 - 4 |
243 |
72 |
BM925547.1 |
- |
3 - 4 |
237 |
72 |
View Full Product Details