Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DNAJC9
Entrez Gene ID
23234
Gene Name
DnaJ heat shock protein family (Hsp40) member C9
Gene Aliases
HDJC9, JDD1, SB73
Chromosome Location
Chr.10: 73241954 - 73247331 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015190.4 NP_056005.1 2 - 3 470 86
GenBank mRNA AF327347.1 - 2 - 3 377 86
AK074420.1 - 2 - 3 406 86
AK094162.1 - 2 - 3 2001 86
BC136507.1 - 2 - 3 397 86

View Full Product Details