The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001018088.2 | NP_001018098.1 | 2 - 3 | 233 | 89 |
NM_017684.4 | NP_060154.3 | 2 - 3 | 233 | 89 | |
NM_018080.3 | NP_060550.2 | 2 - 3 | 233 | 89 | |
NM_020821.2 | NP_065872.1 | 2 - 3 | 233 | 89 | |
XM_011521713.2 | XP_011520015.1 | 2 - 3 | 194 | 89 | |
XM_011521714.2 | XP_011520016.1 | 2 - 3 | 194 | 89 | |
GenBank mRNA | AB037842.1 | - | 2 - 3 | 216 | 89 |
AJ608770.1 | - | 2 - 3 | 216 | 89 | |
AJ608771.1 | - | 2 - 3 | 216 | 89 | |
AJ626860.1 | - | 2 - 3 | 216 | 89 | |
AJ626861.1 | - | 2 - 3 | 216 | 89 | |
AK307518.1 | - | 2 - 3 | 158 | 89 |