The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001018088.2 | NP_001018098.1 | 42 - 43 | 4844 | 153 |
NM_017684.4 | NP_060154.3 | 40 - 41 | 4715 | 153 | |
NM_018080.3 | NP_060550.2 | 40 - 41 | 4715 | 153 | |
NM_020821.2 | NP_065872.1 | 42 - 43 | 4844 | 153 | |
XM_011521713.2 | XP_011520015.1 | 42 - 43 | 4805 | 153 | |
XM_011521714.2 | XP_011520016.1 | 42 - 43 | 4805 | 153 | |
GenBank mRNA | AJ608770.1 | - | 40 - 41 | 4698 | 153 |
AJ608771.1 | - | 42 - 43 | 4827 | 153 | |
AJ626860.1 | - | 40 - 41 | 4698 | 153 | |
AJ626861.1 | - | 42 - 43 | 4827 | 153 |