Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GRHL3
Entrez Gene ID
57822
Gene Name
grainyhead like transcription factor 3
Gene Aliases
SOM, TFCP2L4, VWS2
Chromosome Location
Chr.1: 24319322 - 24364482 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AY231161.1 - 1 - 2 85 110

View Full Product Details