The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_017014867.1 | XP_016870356.1 | 9 - 10 | 985 | 89 |
GenBank mRNA | AK091390.1 | - | 4 - 5 | 1215 | 89 |