Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NOP9
Entrez Gene ID
161424
Gene Name
NOP9 nucleolar protein
Gene Aliases
C14orf21
Chromosome Location
Chr.14: 24299854 - 24309126 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174913.2 NP_777573.1 8 - 9 1751 84
XM_005267385.1 XP_005267442.1 8 - 9 1778 84
XM_011536526.1 XP_011534828.1 8 - 9 1570 84
GenBank mRNA AB095941.1 - 8 - 9 1743 84
AK123717.1 - 3 - 4 526 84
BX248018.1 - 8 - 9 1713 84
BX647311.1 - 8 - 9 1718 84

View Full Product Details