Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IL17RE
Entrez Gene ID
132014
Gene Name
interleukin 17 receptor E
Gene Aliases
-
Chromosome Location
Chr.3: 9902086 - 9916402 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193380.1 NP_001180309.1 1 - 2 237 77
NM_153480.1 NP_705613.1 1 - 2 237 77
NM_153483.2 NP_705616.2 2 - 3 256 77
XM_011533362.2 XP_011531664.1 1 - 2 172 77
XM_011533363.2 XP_011531665.1 1 - 2 172 77
XM_011533364.2 XP_011531666.1 1 - 2 172 77
GenBank mRNA AF458069.1 - 1 - 2 237 77
AK074238.1 - 1 - 2 217 77
AK297697.1 - 1 - 2 203 77
AK314476.1 - 1 - 2 222 77
AY358451.1 - 1 - 2 237 77
BC035478.1 - 2 - 3 249 77
BC063110.1 - 1 - 2 235 77
BC137077.1 - 2 - 3 199 77
BC137078.1 - 2 - 3 199 77

View Full Product Details