The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001007527.1 | NP_001007528.1 | 1 - 2 | 360 | 113 |
XM_017010024.1 | XP_016865513.1 | 1 - 2 | 306 | 113 | |
GenBank mRNA | AK091295.1 | - | 1 - 2 | 300 | 113 |
AK300433.1 | - | 1 - 2 | 356 | 113 | |
AK307575.1 | - | 1 - 2 | 306 | 113 | |
BC110506.2 | - | 1 - 2 | 299 | 113 | |
BX647340.1 | - | 1 - 2 | 335 | 113 | |
CR749399.1 | - | 1 - 2 | 360 | 113 |