The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015079.5 | NP_055894.6 | 11 - 12 | 2574 | 137 |
NM_144572.1 | NP_653173.1 | 11 - 12 | 2574 | 137 | |
XM_011521387.2 | XP_011519689.1 | 10 - 11 | 2519 | 137 | |
XM_017022022.1 | XP_016877511.1 | 13 - 14 | 2595 | 137 | |
GenBank mRNA | AB028978.1 | - | 11 - 12 | 2437 | 137 |
BC033712.1 | - | 6 - 7 | 1357 | 137 | |
BC140766.1 | - | 10 - 11 | 2219 | 137 | |
BC144482.1 | - | 10 - 11 | 2219 | 137 | |
BC152465.1 | - | 11 - 12 | 2437 | 137 |