Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IVNS1ABP
Entrez Gene ID
10625
Gene Name
influenza virus NS1A binding protein
Gene Aliases
FLARA3, HSPC068, KLHL39, ND1, NS-1, NS1-BP, NS1BP
Chromosome Location
Chr.1: 185292367 - 185317343 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006469.4 NP_006460.2 5 - 6 983 131
XM_005244843.3 XP_005244900.1 5 - 6 939 131
XM_011509079.2 XP_011507381.1 5 - 6 993 131
XM_017000084.1 XP_016855573.1 5 - 6 939 131
GenBank mRNA AB020657.1 - 5 - 6 990 131
AF161553.1 - 5 - 6 896 128
AF205218.1 - 5 - 6 915 131
AJ012449.1 - 5 - 6 897 131
AK001625.1 - 3 - 4 321 131
AK023020.1 - 4 - 5 591 131
AK023123.1 - 5 - 6 927 131
AK292431.1 - 5 - 6 733 131
BC067739.1 - 5 - 6 883 131
DQ443528.1 - 5 - 6 872 131

View Full Product Details