The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020309.3 | NP_064705.1 | 7 - 8 | 1042 | 79 |
GenBank mRNA | AB032436.1 | - | 7 - 8 | 891 | 79 |
AK289622.1 | - | 4 - 5 | 450 | 79 | |
AK294226.1 | - | 7 - 8 | 848 | 79 | |
AK294405.1 | - | 7 - 8 | 891 | 79 | |
BC059379.1 | - | 7 - 8 | 1017 | 79 |