The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142463.2 | NP_001135935.1 | 8 - 9 | 997 | 63 |
NM_001288747.1 | NP_001275676.1 | 8 - 9 | 997 | 63 | |
NM_001363.4 | NP_001354.1 | 8 - 9 | 997 | 63 | |
NR_110021.1 | 7 - 8 | 1474 | 63 | ||
NR_110022.1 | 7 - 8 | 1593 | 63 | ||
GenBank mRNA | AF067008.1 | - | 8 - 9 | 833 | 63 |
AK225077.1 | - | 8 - 9 | 861 | 63 | |
AK307672.1 | - | 8 - 9 | 886 | 63 | |
BC009928.2 | - | 8 - 9 | 975 | 63 | |
BC010015.2 | - | 8 - 9 | 799 | 63 | |
JF279874.1 | - | 8 - 9 | 820 | 63 | |
KC954523.1 | - | 7 - 8 | 1297 | 63 | |
KC954524.1 | - | 7 - 8 | 1416 | 63 | |
U59151.1 | - | 8 - 9 | 853 | 63 |