Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- PPP6R2
- Entrez Gene ID
-
9701
- Gene Name
- protein phosphatase 6 regulatory subunit 2
- Gene Aliases
-
KIAA0685, PP6R2, SAP190, SAPS2
- Chromosome Location
-
Chr.22: 50343317 - 50445090 on Build GRCh38
View Full Product Details