Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PPP6R2
Entrez Gene ID
9701
Gene Name
protein phosphatase 6 regulatory subunit 2
Gene Aliases
KIAA0685, PP6R2, SAP190, SAPS2
Chromosome Location
Chr.22: 50343317 - 50445090 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001242898.1 NP_001229827.1 7 - 8 1116 148
NM_001242900.1 NP_001229829.1 7 - 8 1116 148
NM_014678.4 NP_055493.2 7 - 8 1116 148
XM_006724431.2 XP_006724494.1 8 - 9 1227 148
XM_006724434.1 XP_006724497.1 9 - 10 1376 148
XM_006724435.2 XP_006724498.1 7 - 8 1101 148
XM_011530721.2 XP_011529023.1 6 - 7 801 148
XM_011530722.2 XP_011529024.1 6 - 7 801 148
XM_011530730.2 XP_011529032.1 6 - 7 801 148
XM_011530731.1 XP_011529033.1 6 - 7 801 148
XM_017029116.1 XP_016884605.1 6 - 7 801 148
XM_017029120.1 XP_016884609.1 9 - 10 1277 148
XM_017029121.1 XP_016884610.1 7 - 8 1101 148
XM_017029122.1 XP_016884611.1 8 - 9 1179 148
XM_017029123.1 XP_016884612.1 7 - 8 1101 148
XM_017029124.1 XP_016884613.1 8 - 9 1227 148
XM_017029125.1 XP_016884614.1 7 - 8 1101 148
XM_017029127.1 XP_016884616.1 8 - 9 1179 148
XM_017029128.1 XP_016884617.1 8 - 9 1227 148
XM_017029129.1 XP_016884618.1 7 - 8 1101 148
XM_017029130.1 XP_016884619.1 7 - 8 1101 148
XM_017029131.1 XP_016884620.1 8 - 9 1227 148
XM_017029132.1 XP_016884621.1 8 - 9 1179 148
XM_017029134.1 XP_016884623.1 7 - 8 1055 148
XM_017029135.1 XP_016884624.1 8 - 9 1105 148
XM_017029136.1 XP_016884625.1 7 - 8 1055 148
XM_017029137.1 XP_016884626.1 7 - 8 1055 148
XM_017029138.1 XP_016884627.1 7 - 8 1055 148
GenBank mRNA AB014585.1 - 7 - 8 1088 148
AK302472.1 - 7 - 8 1027 148
BC000976.4 - 8 - 9 1105 148
BC006568.2 - 7 - 8 1102 148
BC052995.2 - 7 - 8 1022 148

View Full Product Details