The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080469.1 | NP_001073938.1 | 2 - 2 | 2804 | 108 |
GenBank mRNA | BC046245.1 | - | 1 - 1 | 1696 | 108 |