The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012642.2 | NP_001012660.1 | 6 - 7 | 495 | 143 |
XM_011521327.2 | XP_011519629.1 | 6 - 7 | 604 | 143 | |
XM_011521328.2 | XP_011519630.1 | 6 - 7 | 955 | 143 | |
XM_011521329.2 | XP_011519631.1 | 6 - 7 | 472 | 143 | |
XM_011521330.2 | XP_011519632.1 | 6 - 7 | 556 | 143 | |
XM_011521331.2 | XP_011519633.1 | 6 - 7 | 495 | 143 | |
XM_011521332.2 | XP_011519634.1 | 7 - 8 | 679 | 143 | |
XM_011521333.2 | XP_011519635.1 | 7 - 8 | 696 | 143 | |
XM_017021997.1 | XP_016877486.1 | 6 - 7 | 639 | 143 | |
XM_017021998.1 | XP_016877487.1 | 6 - 7 | 863 | 143 | |
GenBank mRNA | AK095072.1 | - | 6 - 7 | 485 | 143 |
BC038451.1 | - | 6 - 7 | 469 | 143 | |
DA854662.1 | - | 6 - 7 | 495 | 143 |