The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001300734.1 | NP_001287663.1 | 3 - 4 | 651 | 126 |
GenBank mRNA | AB107037.1 | - | 3 - 4 | 456 | 126 |
AK091640.1 | - | 2 - 3 | 1553 | 126 |