Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SNX30
Entrez Gene ID
401548
Gene Name
sorting nexin family member 30
Gene Aliases
ATG24A
Chromosome Location
Chr.9: 112750817 - 112875847 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012994.1 NP_001013012.1 5 - 6 966 93
XM_011518691.2 XP_011516993.1 5 - 6 1003 93
XM_017014716.1 XP_016870205.1 5 - 6 1034 93
GenBank mRNA AK127013.1 - 4 - 5 670 93

View Full Product Details