The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000447.2 | NP_000438.2 | 4 - 5 | 567 | 163 |
NM_012486.2 | NP_036618.2 | 4 - 5 | 567 | 163 | |
XM_005273199.3 | XP_005273256.1 | 3 - 4 | 380 | 163 | |
XM_017001835.1 | XP_016857324.1 | 3 - 4 | 888 | 163 | |
XM_017001836.1 | XP_016857325.1 | 3 - 4 | 380 | 163 | |
GenBank mRNA | AF416718.1 | - | 2 - 3 | 208 | 163 |
AK292299.1 | - | 4 - 6 | 163 | ||
BC006365.2 | - | 4 - 6 | 163 | ||
BT006984.1 | - | 1 - 3 | 163 | ||
DA915823.1 | - | 4 - 5 | 520 | 163 | |
L43964.1 | - | 3 - 4 | 507 | 163 | |
L44577.1 | - | 3 - 5 | 163 | ||
U34349.1 | - | 2 - 3 | 231 | 163 |