The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000447.2 | NP_000438.2 | 8 - 9 | 1220 | 161 |
NM_012486.2 | NP_036618.2 | 8 - 9 | 1220 | 161 | |
XM_005273199.3 | XP_005273256.1 | 7 - 8 | 1033 | 161 | |
XM_017001835.1 | XP_016857324.1 | 7 - 8 | 1541 | 161 | |
XM_017001836.1 | XP_016857325.1 | 7 - 8 | 1033 | 161 | |
GenBank mRNA | AK292299.1 | - | 9 - 10 | 1172 | 161 |
AK301143.1 | - | 7 - 8 | 969 | 161 | |
BC006365.2 | - | 9 - 10 | 1414 | 161 | |
BT006984.1 | - | 6 - 7 | 793 | 161 | |
L43964.1 | - | 7 - 8 | 1160 | 161 | |
L44577.1 | - | 8 - 9 | 1158 | 161 | |
U34349.1 | - | 6 - 7 | 884 | 161 |