The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_139069.2 | NP_620708.1 | 7 - 8 | 956 | 131 |
NM_139070.2 | NP_620709.1 | 7 - 8 | 956 | 131 | |
XM_017009638.1 | XP_016865127.1 | 8 - 9 | 925 | 131 | |
XM_017009640.1 | XP_016865129.1 | 8 - 9 | 1073 | 131 | |
GenBank mRNA | AK289638.1 | - | 7 - 8 | 852 | 131 |
HM036708.1 | - | 6 - 7 | 705 | 131 | |
U35002.1 | - | 6 - 7 | 707 | 131 | |
U35003.1 | - | 6 - 7 | 719 | 131 |