The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022170.1 | NP_071496.1 | 3 - 4 | 318 | 136 |
NM_031992.1 | NP_114381.1 | 3 - 4 | 318 | 136 | |
XM_011516556.1 | XP_011514858.1 | 3 - 4 | 351 | 136 | |
XM_011516557.1 | XP_011514859.1 | 3 - 4 | 336 | 136 | |
GenBank mRNA | AK290676.1 | - | 3 - 4 | 338 | 136 |
AK297651.1 | - | 3 - 4 | 363 | 136 | |
AK314749.1 | - | 3 - 4 | 420 | 136 | |
BC010021.1 | - | 1 - 1 | 316 | 136 | |
BC021214.2 | - | 3 - 4 | 316 | 136 | |
BC066928.1 | - | 1 - 1 | 335 | 136 | |
D26068.1 | - | 3 - 4 | 318 | 136 |