The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001143838.2 | NP_001137310.1 | 3 - 4 | 458 | 178 |
NM_001284510.1 | NP_001271439.1 | 2 - 3 | 329 | 178 | |
NM_177550.4 | NP_808218.1 | 3 - 4 | 458 | 178 | |
XM_011523795.2 | XP_011522097.1 | 3 - 4 | 448 | 178 | |
GenBank mRNA | AJ489980.1 | - | 2 - 3 | 241 | 178 |
AK172785.1 | - | 3 - 4 | 403 | 178 | |
AK297612.1 | - | 2 - 3 | 274 | 178 | |
AY151833.1 | - | 3 - 4 | 382 | 178 | |
BC104795.1 | - | 3 - 4 | 414 | 178 | |
BC112151.1 | - | 3 - 4 | 414 | 178 | |
BC143689.1 | - | 3 - 4 | 414 | 178 |