The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001143838.2 | NP_001137310.1 | 8 - 9 | 1242 | 106 |
NM_001284509.1 | NP_001271438.1 | 8 - 9 | 1191 | 106 | |
NM_001284510.1 | NP_001271439.1 | 7 - 8 | 1113 | 106 | |
NM_177550.4 | NP_808218.1 | 8 - 9 | 1242 | 106 | |
XM_011523795.2 | XP_011522097.1 | 8 - 9 | 1232 | 106 | |
GenBank mRNA | AK127797.1 | - | 8 - 9 | 1139 | 106 |
AK172779.1 | - | 1 - 2 | 95 | 106 | |
AK297612.1 | - | 7 - 8 | 1058 | 106 | |
AY151833.1 | - | 8 - 9 | 1166 | 106 | |
BC104795.1 | - | 8 - 9 | 1198 | 106 | |
BC112151.1 | - | 8 - 9 | 1198 | 106 | |
BC143689.1 | - | 8 - 9 | 1198 | 106 | |
CR749584.1 | - | 3 - 4 | 356 | 106 |