Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC13A5
Entrez Gene ID
284111
Gene Name
solute carrier family 13 member 5
Gene Aliases
EIEE25, NACT, mIndy
Chromosome Location
Chr.17: 6684713 - 6713421 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001143838.2 NP_001137310.1 9 - 10 1361 170
NM_001284509.1 NP_001271438.1 9 - 10 1310 170
NM_001284510.1 NP_001271439.1 8 - 9 1232 170
NM_177550.4 NP_808218.1 9 - 10 1361 170
XM_011523795.2 XP_011522097.1 9 - 10 1351 170
GenBank mRNA AK127797.1 - 9 - 10 1258 170
AK172779.1 - 2 - 3 214 170
AK172785.1 - 9 - 10 1306 170
AK297612.1 - 8 - 9 1177 170
AY151833.1 - 9 - 10 1285 170
BC104795.1 - 9 - 10 1317 170
BC112151.1 - 9 - 10 1317 170
BC143689.1 - 9 - 10 1317 170
CR749584.1 - 4 - 5 475 170

View Full Product Details