The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270546.1 | NP_001257475.1 | 21 - 22 | 3099 | 148 |
NM_006738.5 | NP_006729.4 | 29 - 30 | 7300 | 148 | |
NM_007200.4 | NP_009131.2 | 29 - 30 | 7288 | 148 | |
GenBank mRNA | AB055890.1 | - | 30 - 31 | 7307 | 148 |
AB209414.1 | - | 18 - 19 | 2581 | 148 | |
AF126008.1 | - | 21 - 22 | 2942 | 148 | |
AF387101.1 | - | 28 - 29 | 7094 | 148 | |
AF406992.1 | - | 29 - 30 | 7187 | 148 | |
BC063592.1 | - | 16 - 17 | 2075 | 148 | |
BC140862.1 | - | 14 - 15 | 1911 | 148 | |
BC171798.1 | - | 14 - 15 | 1911 | 148 |