Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ASPH
Entrez Gene ID
444
Gene Name
aspartate beta-hydroxylase
Gene Aliases
AAH, BAH, CASQ2BP1, FDLAB, HAAH, JCTN, junctin
Chromosome Location
Chr.8: 61500556 - 61714640 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164754.1 NP_001158226.1 1 - 2 374 170
NM_001164755.1 NP_001158227.1 1 - 2 374 170
NM_001164756.1 NP_001158228.1 1 - 2 374 170
NM_004318.3 NP_004309.2 1 - 2 374 170
NM_032466.3 NP_115855.1 1 - 2 374 170
XM_005251235.2 XP_005251292.1 1 - 2 343 170
XM_005251236.2 XP_005251293.1 1 - 2 343 170
XM_005251238.2 XP_005251295.1 1 - 2 343 170
XM_005251239.2 XP_005251296.1 1 - 2 343 170
XM_005251242.2 XP_005251299.1 1 - 2 343 170
XM_005251243.2 XP_005251300.1 1 - 2 343 170
XM_005251246.2 XP_005251303.1 1 - 2 343 170
XM_005251247.2 XP_005251304.1 1 - 2 343 170
XM_005251250.2 XP_005251307.1 1 - 2 343 170
XM_017013419.1 XP_016868908.1 1 - 2 343 170
XM_017013420.1 XP_016868909.1 1 - 2 343 170
XM_017013421.1 XP_016868910.1 1 - 2 343 170
XM_017013422.1 XP_016868911.1 1 - 2 343 170
XM_017013423.1 XP_016868912.1 1 - 2 343 170
XM_017013424.1 XP_016868913.1 1 - 2 343 170
XM_017013425.1 XP_016868914.1 1 - 2 343 170
XM_017013428.1 XP_016868917.1 1 - 2 343 170
XM_017013429.1 XP_016868918.1 1 - 2 343 170
XM_017013430.1 XP_016868919.1 1 - 2 343 170
XM_017013432.1 XP_016868921.1 1 - 2 343 170
XM_017013434.1 XP_016868923.1 1 - 2 343 170
XM_017013436.1 XP_016868925.1 1 - 2 343 170
XM_017013437.1 XP_016868926.1 1 - 2 343 170
XM_017013438.1 XP_016868927.1 1 - 2 343 170
XM_017013441.1 XP_016868930.1 1 - 2 343 170
XM_017013442.1 XP_016868931.1 1 - 2 343 170
XM_017013443.1 XP_016868932.1 1 - 2 343 170
GenBank mRNA AF289489.1 - 1 - 2 116 170
AK291836.1 - 1 - 2 187 170
AK298577.1 - 2 - 3 295 143
BC015518.2 - 1 - 2 293 170
BC025236.1 - 1 - 2 302 170
BC066929.1 - 1 - 2 270 170
BC142967.1 - 1 - 2 251 170
CR996516.1 - 1 - 2 374 170
DA136318.1 - 1 - 2 343 170
EU718494.1 - 1 - 2 182 170
U03109.1 - 1 - 2 182 170

View Full Product Details