The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001193431.2 | NP_001180360.1 | 10 - 11 | 970 | 140 |
NM_001308297.1 | NP_001295226.1 | 9 - 10 | 898 | 140 | |
NM_015967.6 | NP_057051.3 | 10 - 11 | 970 | 140 | |
XM_011541222.1 | XP_011539524.1 | 10 - 11 | 951 | 140 | |
XM_011541223.2 | XP_011539525.1 | 10 - 11 | 951 | 140 | |
XM_011541225.2 | XP_011539527.1 | 9 - 10 | 900 | 140 | |
XM_017001004.1 | XP_016856493.1 | 10 - 11 | 951 | 140 | |
XM_017001005.1 | XP_016856494.1 | 10 - 11 | 932 | 140 | |
GenBank mRNA | AF001846.1 | - | 10 - 11 | 878 | 140 |
AF077031.1 | - | 10 - 11 | 906 | 140 | |
AF150732.1 | - | 10 - 11 | 904 | 140 | |
AK303124.1 | - | 6 - 7 | 605 | 140 | |
AK310570.1 | - | 5 - 6 | 542 | 140 | |
AK310698.1 | - | 10 - 11 | 923 | 140 | |
GU479452.1 | - | 10 - 11 | 884 | 140 | |
JN084012.1 | - | 9 - 10 | 775 | 140 |