The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080429.2 | NP_536354.2 | 4 - 5 | 527 | 103 |
XM_011510104.2 | XP_011508406.1 | 4 - 5 | 587 | 103 | |
GenBank mRNA | AB066105.1 | - | 4 - 5 | 527 | 103 |
AF159174.1 | - | 4 - 5 | 523 | 103 | |
AK314993.1 | - | 4 - 5 | 550 | 103 | |
BC069607.1 | - | 4 - 5 | 527 | 103 | |
BC074896.2 | - | 4 - 5 | 524 | 103 | |
BC074897.2 | - | 4 - 5 | 524 | 103 |