The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_181787.2 | NP_861452.2 | 14 - 15 | 1700 | 149 |
XM_005250894.4 | XP_005250951.2 | 14 - 15 | 1913 | 149 | |
XM_005250895.3 | XP_005250952.1 | 13 - 14 | 1581 | 149 | |
XM_011516983.2 | XP_011515285.1 | 13 - 14 | 1607 | 149 | |
XM_011516984.2 | XP_011515286.1 | 13 - 14 | 1580 | 149 | |
XM_017013351.1 | XP_016868840.1 | 13 - 14 | 1574 | 149 | |
XM_017013352.1 | XP_016868841.1 | 12 - 13 | 1651 | 149 | |
XM_017013353.1 | XP_016868842.1 | 12 - 13 | 1475 | 149 | |
XM_017013354.1 | XP_016868843.1 | 13 - 14 | 1606 | 149 | |
XM_017013355.1 | XP_016868844.1 | 13 - 14 | 1599 | 149 | |
XM_017013356.1 | XP_016868845.1 | 12 - 13 | 1452 | 149 | |
XM_017013357.1 | XP_016868846.1 | 12 - 13 | 1440 | 149 | |
GenBank mRNA | AK123618.1 | - | 6 - 7 | 695 | 149 |
AK123682.1 | - | 14 - 15 | 1654 | 149 | |
AL832859.1 | - | 8 - 9 | 901 | 149 | |
BC110870.1 | - | 14 - 15 | 1628 | 149 | |
BC126193.1 | - | 14 - 15 | 1596 | 149 | |
BC130576.1 | - | 14 - 15 | 1596 | 149 | |
BX538048.1 | - | 14 - 15 | 1648 | 149 | |
BX538174.1 | - | 14 - 15 | 1652 | 149 |