The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_181787.2 | NP_861452.2 | 8 - 9 | 991 | 151 |
XM_005250894.4 | XP_005250951.2 | 8 - 9 | 1204 | 151 | |
XM_005250895.3 | XP_005250952.1 | 7 - 8 | 872 | 151 | |
XM_011516983.2 | XP_011515285.1 | 8 - 9 | 997 | 151 | |
XM_011516984.2 | XP_011515286.1 | 7 - 8 | 871 | 151 | |
XM_017013352.1 | XP_016868841.1 | 6 - 7 | 942 | 151 | |
XM_017013354.1 | XP_016868843.1 | 7 - 8 | 897 | 151 | |
XM_017013355.1 | XP_016868844.1 | 7 - 8 | 890 | 151 | |
XM_017013356.1 | XP_016868845.1 | 6 - 7 | 743 | 151 | |
GenBank mRNA | AK123682.1 | - | 8 - 9 | 945 | 151 |
AL832859.1 | - | 3 - 4 | 306 | 151 | |
BC110870.1 | - | 8 - 9 | 919 | 151 | |
BC126193.1 | - | 8 - 9 | 887 | 151 | |
BC130576.1 | - | 8 - 9 | 887 | 151 | |
BX538048.1 | - | 8 - 9 | 939 | 151 | |
BX538174.1 | - | 8 - 9 | 943 | 151 |