The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001173988.1 | NP_001167459.1 | 13 - 14 | 2210 | 129 |
NM_024718.4 | NP_078994.3 | 13 - 14 | 2207 | 129 | |
GenBank mRNA | AK027586.1 | - | 10 - 11 | 1628 | 129 |
AK128154.2 | - | 13 - 14 | 2471 | 129 | |
AL713707.1 | - | 6 - 7 | 1183 | 129 | |
BC021095.1 | - | 13 - 14 | 1911 | 129 | |
BC024964.2 | - | 4 - 5 | 693 | 129 | |
BC035786.1 | - | 13 - 14 | 2165 | 129 | |
DQ141240.1 | - | 13 - 14 | 1932 | 129 | |
EF156752.1 | - | 13 - 14 | 1934 | 129 |