Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TDRD5
Entrez Gene ID
163589
Gene Name
tudor domain containing 5
Gene Aliases
TUDOR3
Chromosome Location
Chr.1: 179591613 - 179691272 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199085.1 NP_001186014.1 16 - 17 3163 153
NM_001199089.1 NP_001186018.1 16 - 17 2876 153
NM_001199091.1 NP_001186020.1 15 - 16 2842 153
NM_001199092.1 NP_001186021.1 12 - 13 1686 153
NM_173533.3 NP_775804.2 15 - 16 3001 153
XM_005244934.1 XP_005244991.1 16 - 17 2948 153
XM_005244935.4 XP_005244992.1 16 - 17 2853 153
XM_017000473.1 XP_016855962.1 15 - 16 2707 153
GenBank mRNA AK092142.1 - 12 - 13 1698 152
AK126889.1 - 15 - 16 2842 153
BC043368.1 - 7 - 8 1013 153
BC089387.1 - 13 - 14 1903 153
BC130532.1 - 16 - 17 2840 153
BC144058.1 - 16 - 17 2895 153

View Full Product Details