Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TDRD5
Entrez Gene ID
163589
Gene Name
tudor domain containing 5
Gene Aliases
TUDOR3
Chromosome Location
Chr.1: 179591613 - 179691272 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199085.1 NP_001186014.1 17 - 18 3378 129
NM_001199089.1 NP_001186018.1 17 - 18 3091 129
NM_001199091.1 NP_001186020.1 16 - 17 3057 129
NM_001199092.1 NP_001186021.1 13 - 14 1901 129
NM_173533.3 NP_775804.2 16 - 17 3216 129
XM_005244934.1 XP_005244991.1 17 - 18 3163 129
XM_005244935.4 XP_005244992.1 17 - 18 3068 129
XM_017000473.1 XP_016855962.1 16 - 17 2922 129
GenBank mRNA AK092142.1 - 13 - 14 1913 129
AK126889.1 - 16 - 17 3057 129
AK127370.1 - 16 - 17 2948 129
BC043368.1 - 8 - 9 1228 129
BC089387.1 - 14 - 15 2118 129
BC130532.1 - 17 - 18 3055 129
BC144058.1 - 17 - 18 3110 129

View Full Product Details