Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TDRD5
Entrez Gene ID
163589
Gene Name
tudor domain containing 5
Gene Aliases
TUDOR3
Chromosome Location
Chr.1: 179591613 - 179691272 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199085.1 NP_001186014.1 12 - 13 2525 84
NM_001199089.1 NP_001186018.1 12 - 13 2238 84
NM_001199091.1 NP_001186020.1 12 - 13 2366 84
NM_001199092.1 NP_001186021.1 9 - 10 1210 84
NM_173533.3 NP_775804.2 12 - 13 2525 84
XM_005244934.1 XP_005244991.1 12 - 13 2310 84
XM_005244935.4 XP_005244992.1 12 - 13 2215 84
XM_017000473.1 XP_016855962.1 12 - 13 2231 84
GenBank mRNA AK092142.1 - 9 - 10 1223 84
AK126889.1 - 12 - 13 2366 84
AK127370.1 - 12 - 13 2257 84
BC043368.1 - 3 - 4 375 84
BC065707.1 - 9 - 10 1334 84
BC089387.1 - 10 - 11 1427 84
BC130532.1 - 12 - 13 2202 84
BC144058.1 - 12 - 13 2257 84

View Full Product Details