The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017767.2 | NP_060237.2 | 4 - 5 | 1220 | 153 |
NM_130849.3 | NP_570901.2 | 5 - 6 | 1076 | 153 | |
XM_006716599.1 | XP_006716662.1 | 5 - 6 | 1055 | 153 | |
GenBank mRNA | AK000695.1 | - | 4 - 5 | 1135 | 153 |
AK025537.1 | - | 5 - 6 | 1070 | 153 | |
AK290563.1 | - | 5 - 6 | 1024 | 153 | |
BC062625.1 | - | 5 - 6 | 1056 | 153 | |
KU178707.1 | - | 5 - 6 | 970 | 153 | |
KU178708.1 | - | 4 - 5 | 688 | 153 |