The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040025.1 | NP_001035114.1 | 2 - 3 | 292 | 120 |
GenBank mRNA | AK303526.1 | - | 2 - 3 | 121 | 120 |
AK304718.1 | - | 2 - 3 | 131 | 120 | |
BC042380.1 | - | 2 - 3 | 130 | 120 | |
BC063687.1 | - | 2 - 3 | 94 | 120 | |
BC105076.1 | - | 2 - 3 | 292 | 120 | |
BC105078.1 | - | 2 - 3 | 292 | 120 |