Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TDRD5
Entrez Gene ID
163589
Gene Name
tudor domain containing 5
Gene Aliases
TUDOR3
Chromosome Location
Chr.1: 179591613 - 179691272 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199085.1 NP_001186014.1 3 - 4 1158 97
NM_001199089.1 NP_001186018.1 3 - 4 871 97
NM_001199091.1 NP_001186020.1 3 - 4 999 97
NM_173533.3 NP_775804.2 3 - 4 1158 97
XM_005244934.1 XP_005244991.1 3 - 4 943 97
XM_005244935.4 XP_005244992.1 3 - 4 848 97
XM_017000473.1 XP_016855962.1 3 - 4 864 97
GenBank mRNA AK126889.1 - 3 - 4 998 97
AK127370.1 - 3 - 4 890 97
BC089387.1 - 1 - 2 60 97
BC130532.1 - 3 - 4 835 97
BC144058.1 - 3 - 4 890 97

View Full Product Details