Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLC13A5
Entrez Gene ID
284111
Gene Name
solute carrier family 13 member 5
Gene Aliases
EIEE25, NACT, mIndy
Chromosome Location
Chr.17: 6684713 - 6713421 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001143838.2 NP_001137310.1 2 - 3 319 73
NM_001284509.1 NP_001271438.1 2 - 3 319 73
NM_177550.4 NP_808218.1 2 - 3 319 73
XM_011523795.2 XP_011522097.1 2 - 3 309 73
GenBank mRNA AK127797.1 - 2 - 3 267 73
AK172785.1 - 2 - 3 264 73
AY151833.1 - 2 - 3 243 73
BC104795.1 - 2 - 3 275 73
BC112151.1 - 2 - 3 275 73
BC143689.1 - 2 - 3 275 73
DA187094.1 - 2 - 3 319 73

View Full Product Details